4-Ethoxyphenol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
B20584.09
B20584.18
B20584.09EA
1200
CZK
B20584.09
B20584.18
4-Ethoxyphenol ≥99%
4-Ethoxyphenol
Purity: 99%
CAS No.: 622-62-8
Molecular Formula: C8H10O2
Formula Weight: 138.17
Odour: Phenol-like
Melting Point: 64-67°
Boiling Point: 121°/9mm
Beilstein Reference: 1907114
MDL No.: MFCD00002334
Vzorec:
C₈H₁₀O₂ MW: 138,17 g/mol Bod varu: 121 °C (9 mmHg) Bod tání: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002334 CAS číslo: 622-62-8 EINECS: 210-748-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...