Anizol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Purity: 99%
CAS No.: 100-66-3
Molecular Formula: C7H8O
Formula Weight: 108.14
Flash Point: 43°(109°F)
Refractive Index: 1.5170
Odour: Ether-like
Melting Point: -37°
Boiling Point: 153-154°
Beilstein Reference: 506892
Merck Reference: 14,669
UN No.: 2222
MDL No.: MFCD00008354
Vzorec:
C₇H₈O Bod varu: 153…154 °C Bod tání: –37 °C Hustota: 0,994 Bod vzplanutí: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008354 CAS číslo: 100-66-3 EINECS: 202-876-1 UN: 2222 ADR: 3,III Merck Index: 14,669 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...