Tisk…

Triphenylamine 98%

Dodavatel: Thermo Fisher Scientific
Varování

A15939.06 A15939.14 A15939.22
A15939.06EA 824 CZK
A15939.06 A15939.14 A15939.22
Triphenylamine 98%
Triphenylamine

Purity: 98% CAS No.: 603-34-9 Molecular Formula: (C6H5)3N Formula Weight: 245.33 Melting Point: 125-129° Boiling Point: 365° MDL No.: MFCD00003020

Vzorec: C₁₈H₁₅N
Bod varu: 365 °C
Bod tání: 125…129 °C
Hustota: 0,774 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003020
CAS číslo: 603-34-9
EINECS: 210-035-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR