Tisk…

2-Nitroanisole ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A19518.0E A19518.36
A19518.0EEA 7760 CZK
A19518.0E A19518.36
2-Nitroanisole ≥99%
2-Nitroanisole

Purity: 99% CAS No.: 91-23-6 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 142°(287°F) Refractive Index: 1.5610 Melting Point: 9-10° Boiling Point: 272-273° Beilstein Reference: 1868032 Merck Reference: 14,6585 UN No.: 2730 MDL No.: MFCD00007096

Vzorec: C₇H₇NO₃
MW: 153,14 g/mol
Bod varu: 272…273 °C
Bod tání: 9…10 °C
Hustota: 1,254
Bod vzplanutí: 142 °C (287 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007096
CAS číslo: 91-23-6
EINECS: 202-052-1
UN: 2730
ADR: 6.1,III
Merck Index: 14,6585

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR