Tisk…

Nitrobenzen ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A10585.0C A10585.0E A10585.36
A10585.0CEA 5840 CZK
A10585.0C A10585.0E A10585.36
Nitrobenzen ≥99%
Nitrobenzen

Purity: 99% CAS No.: 98-95-3 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 87°(188°F) Refractive Index: 1.5520 Melting Point: 5-6° Boiling Point: 210-212° Beilstein Reference: 507540 Merck Reference: 14,6588 UN No.: 1662 MDL No.: MFCD00007043

Vzorec: C₆H₅NO₂
Bod varu: 210…212 °C
Bod tání: 5…6 °C
Hustota: 1,196
Bod vzplanutí: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS číslo: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1,II
Merck Index: 14,6588

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR