Tisk…

N-Methyl-2-nitroaniline ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A19538.22 A19538.14 A19538.06
A19538.22EA 9540 CZK
A19538.22 A19538.14 A19538.06
N-Methyl-2-nitroaniline ≥98%
N-Methyl-2-nitroaniline

Purity: 98% CAS No.: 612-28-2 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Flash Point: 110°(230°F) Odour: Characteristic UN No.: 2811 MDL No.: MFCD00007090

Vzorec: C₇H₈N₂O₂
Bod varu: 158 °C (18 mmHg)
Bod tání: 33…36 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007090
CAS číslo: 612-28-2
EINECS: 210-303-1
UN: 2811
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR