N-Butylaniline ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
B24180.14
B24180.22
B24180.14EA
1630
CZK
B24180.14
B24180.22
N-Butylaniline ≥99%
N-Butylaniline
Purity: 99%
CAS No.: 1126-78-9
Molecular Formula: C10H15N
Formula Weight: 149.23
Flash Point: 105°(222°F)
Refractive Index: 1.5320
Boiling Point: 240-242°
UN No.: 2738
MDL No.: MFCD00035798
Vzorec:
(C₆H₅)NHCH₂CH₂CH₂CH₃ MW: 149,24 g/mol Bod varu: 240…242 °C Bod tání: –14,4 °C Hustota: 0,932 Bod vzplanutí: 105 °C (222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00035798 CAS číslo: 1126-78-9 EINECS: 214-425-6 UN: 2738 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...