3,4,5-Trimethoxyaniline ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L01619.06
L01619.14
L01619.06EA
1120
CZK
L01619.06
L01619.14
3,4,5-Trimethoxyaniline ≥98%
3,4,5-Trimethoxyaniline
Purity: 98+%
CAS No.: 24313-88-0
Molecular Formula: C9H13NO3
Formula Weight: 183.21
Melting Point: 110-113°
Beilstein Reference: 642919
UN No.: 2811
MDL No.: MFCD00008393
Vzorec:
C₉H₁₃NO₃ MW: 183,21 g/mol Bod varu: 293 °C (1013 hPa) Bod tání: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008393 CAS číslo: 24313-88-0 EINECS: 246-154-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...