3,5-Dimethoxyaniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15073.09
A15073.14
A15073.18
A15073.09EA
1980
CZK
A15073.09
A15073.14
A15073.18
3,5-Dimethoxyaniline ≥98%
3,5-Dimethoxyaniline
Purity: 98%
CAS No.: 10272-07-8
Molecular Formula: C8H11NO2
Formula Weight: 153.18
Flash Point:
Vzorec:
C₈H₁₁NO₂ Bod varu: 178 °C (20 mmHg) Bod tání: 53…57 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008392 CAS číslo: 10272-07-8 EINECS: 233-616-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...