Tisk…

2,5-Dimethoxyaniline ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

L05053.18 L05053.30
L05053.18EA 1230 CZK
L05053.18 L05053.30
2,5-Dimethoxyaniline ≥99%
2,5-Dimethoxyaniline

Purity: 99% CAS No.: 102-56-7 Molecular Formula: C8H11NO2 Formula Weight: 153.18 Flash Point: 130°(266°F) Odour: Characteristic Boiling Point: 140°/7mm Beilstein Reference: 776823 UN No.: 2811 MDL No.: MFCD00008368

Vzorec: C₈H₁₁NO₂
MW: 153,18 g/mol
Bod varu: 140 °C (7 mmHg)
Bod tání: 78…82 °C
Bod vzplanutí: 130 °C (266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008368
CAS číslo: 102-56-7
EINECS: 203-040-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR