Tisk…
Tissue and biopsy cassettes in QuickLoad™ stacks, Swingsette™, M517T
Dodavatel: Simport Scientific
M517-4T M517-6T M517-2T M517-10T M517-11T M517-12T M517-3T M517-5T M517-7T M517-8T M517-9T
720-1982EA 14080 CZK
720-1982 720-1983 SIMPM517-2T SIMPM517-10T SIMPM517-11T SIMPM517-12T SIMPM517-3T SIMPM517-5T SIMPM517-7T SIMPM517-8T SIMPM517-9T
Tissue and biopsy cassettes in QuickLoad™ stacks, Swingsette™, M517T
Kazety pro zalévání vzorků Kazety tkáňové
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage. Suitable for use with Leica and Sakura labelers.
  • Made of acetal
  • Taped cassettes to be used with Leica and Sakura Ink Jet printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge