2-Nitroanilin ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
o-Nitroaniline
A11055.0B
A11055.0I
A11055.30
A11055.0BEA
2750
CZK
A11055.0B
A11055.0I
A11055.30
2-Nitroanilin ≥98%
2-Nitroanilin
Purity: 98%
CAS No.: 88-74-4
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Flash Point: 168°(334°F)
Melting Point: 70-74°
Boiling Point: 284-286°
Beilstein Reference: 509275
Merck Reference: 14,6583
UN No.: 1661
MDL No.: MFCD00007687
Vzorec:
C₆H₆N₂O₂ MW: 138,13 g/mol Bod varu: 284…286 °C Bod tání: 70…74 °C Hustota: 1,255 Bod vzplanutí: 168 °C (334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007687 CAS číslo: 88-74-4 EINECS: 201-855-4 UN: 1661 ADR: 6.1,II Merck Index: 14,6583 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...