Anilin 99+% ACS
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Aminobenzene, Phenylamine
423420010
423420050
423425000
423420025
ACRO423420010EA
5920
CZK
ACRO423420010
ACRO423420050
ACRO423425000
ACRO423420025
Anilin 99+% ACS
Aniline
Vzorec:
C₆H₅NH₂ MW: 93,13 g/mol Bod varu: 184 °C (1013 hPa) Bod tání: –6,2 °C Hustota: 1,0213 g/cm³ Bod vzplanutí: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS číslo: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1,II Merck Index: 13,00661 |
Výsledky specifikačního testu
Appearance | Clear liquid (May darken to red-brown on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5845 to 1.5875 (20°C, 589 nm) |
GC | ≥99.0 % |
Color scale | ≤250 APHA |
Residue after ignition | ≤0.005 % |
Hydrocarbons | Passes test |
Chlorobenzene (C6H5Cl) | ≤0.01 % |
Nitrobenzene (C6H5NO2) | (Limit about 0.001 %) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...