Anilin 99.5%, extra pure
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aminobenzene, Phenylamine
221730010
221732500
221730025
221730050
ACRO221730010EA
1850
CZK
ACRO221730010
ACRO221732500
ACRO221730025
ACRO221730050
Anilin 99.5%, extra pure
Anilin
Vzorec:
C₆H₅NH₂ MW: 93.13 Bod varu: 181…185 °C (760 mmHg) Bod tání: –6.2 °C Hustota: 1.0213 g/cm³ Bod vzplanutí: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS číslo: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1 II Merck Index: 13,00661 |
|
Výsledky specifikačního testu
Appearance | Clear colorless to yellow to orange to light brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5845 to 1.5875 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...