N,N-Dimethyl-3-nitroaniline ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L00401.06
L00401.14
L00401.06EA
889
CZK
L00401.06
L00401.14
N,N-Dimethyl-3-nitroaniline ≥98%
N,N-Dimethyl-3-nitroaniline
Purity: 98%
CAS No.: 619-31-8
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 57-60°
Beilstein Reference: 1841745
UN No.: 2811
MDL No.: MFCD00007236
Vzorec:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Bod varu: 280…285 °C (1013 hPa) Bod tání: 57…60 °C Hustota: 1,313 g/cm³ (17 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007236 CAS číslo: 619-31-8 EINECS: 210-590-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...