1-Iodo-3-nitrobenzene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B21605.14
B21605.22
B21605.14EA
1560
CZK
B21605.14
B21605.22
1-Iodo-3-nitrobenzene ≥99%
1-Iodo-3-nitrobenzene
Purity: 99%
CAS No.: 645-00-1
Molecular Formula: C6H4INO2
Formula Weight: 249.01
Flash Point: 71°(159°F)
Melting Point: 35-38°
Boiling Point: 279-280°
Beilstein Reference: 1525167
UN No.: 2926
MDL No.: MFCD00007218
Vzorec:
C₆H₄INO₂ MW: 249,01 g/mol Bod varu: 279…280 °C Bod tání: 35…38 °C Hustota: 2,227 g/cm³ (20 °C) Bod vzplanutí: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007218 CAS číslo: 645-00-1 EINECS: 211-427-9 UN: 2926 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...