Tisk…

2,3,5-Triiodobenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: TIBA

L02679.06 L02679.14 L02679.22
L02679.06EA 1350 CZK
L02679.06 L02679.14 L02679.22
2,3,5-Triiodobenzoic acid ≥98%
2,3,5-Triiodobenzoic acid

Purity: 98+% CAS No.: 88-82-4 Molecular Formula: C7H3I3O2 Formula Weight: 499.81 Melting Point: 220-222° Beilstein Reference: 1955088

Vzorec: C₇H₃I₃O₂
MW: 499,81 g/mol
Bod varu: 416 °C (1013 hPa)
Bod tání: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00002420
CAS číslo: 88-82-4
EINECS: 201-859-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR