4-Iodobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
p-Iodobenzonitrile
B25104.06
B25104.14
B25104.22
B25104.06EA
2290
CZK
B25104.06
B25104.14
B25104.22
4-Iodobenzonitrile ≥98%
4-Iodobenzonitrile
Purity: 98%
CAS No.: 3058-39-7
Molecular Formula: C7H4IN
Formula Weight: 229.02
Melting Point: 125-127°
UN No.: 3439
MDL No.: MFCD00051310
Vzorec:
C₇H₄IN MW: 229,02 g/mol Bod varu: 285 °C (1013 hPa) Bod tání: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051310 CAS číslo: 3058-39-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...