Tisk…

5-Iodovanillin ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

A15162.09 A15162.18 A15162.30
A15162.09EA 999 CZK
A15162.09 A15162.18 A15162.30
5-Iodovanillin ≥98%
5-Iodovanillin

Purity: 98% CAS No.: 5438-36-8 Molecular Formula: C8H7IO3 Formula Weight: 278.05 Odour: Irritating Melting Point: 181-184° Beilstein Reference: 2049762 MDL No.: MFCD00006941

Vzorec: C₈H₇IO₃
Bod varu: 335 °C (1013 hPa)
Bod tání: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006941
CAS číslo: 5438-36-8
EINECS: 226-617-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR