Tisk…

2-Iodobenzaldehyde ≥98%

Dodavatel: Thermo Scientific
Varování

L17446.03 L17446.06
L17446.03EA 3110 CZK
L17446.03 L17446.06
2-Iodobenzaldehyde ≥98%
2-Iodobenzaldehyde

Purity: 98% CAS No.: 26260-02-6 Molecular Formula: C7H5IO Formula Weight: 232.02 Storage Temperature: 2°C to 8°C Melting Point: 36-37° Boiling Point: 102°/3mm Beilstein Reference: 1927296 MDL No.: MFCD00039570

Vzorec: C₇H₅IO
MW: 232,02 g/mol
Bod varu: 102 °C (3 mmHg)
Bod tání: 36…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039570
CAS číslo: 26260-02-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR