Tisk…

3-Fluorothiophenol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Fluorobenzene-1-thiol, 1-Fluoro-3-mercaptobenzene, m-Fluorothiophenol, 3-Fluorobenzenethiol

B23114.03 B23114.06 B23114.14
B23114.03EA 1030 CZK
B23114.03 B23114.06 B23114.14
3-Fluorothiophenol ≥98%
3-Fluorothiophenol

Purity: 98% CAS No.: 2557-77-9 Molecular Formula: C6H5FS Formula Weight: 128.17 Flash Point: 58°(136°F) Refractive Index: 1.5540 Odour: Stench Boiling Point: 156° Beilstein Reference: 2039302 UN No.: 3071 MDL No.: MFCD00040227

Vzorec: C₆H₅FS
MW: 128.17
Bod varu: 156 °C
Hustota: 1.18
Bod vzplanutí: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040227
CAS číslo: 2557-77-9
EINECS: 219-876-2
UN: 3071
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR