Tisk…

2-Fluorothiophenol ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Fluorobenzenethiol, 1-Fluoro-2-mercaptobenzene, 2-Fluorobenzene-1-thiol, o-Fluorothiophenol

B24995.03 B24995.06 B24995.14
B24995.03EA 714 CZK
B24995.03 B24995.06 B24995.14
2-Fluorothiophenol ≥97%
2-Fluorothiophenol

Purity: 97% CAS No.: 2557-78-0 Molecular Formula: C6H5FS Formula Weight: 128.17 Flash Point: 56°(133°F) Refractive Index: 1.5550 Odour: Stench Boiling Point: 164° Beilstein Reference: 2039768 UN No.: 3071 MDL No.: MFCD00041419

Vzorec: C₆H₅FS
MW: 128.17
Bod varu: 164 °C
Hustota: 1.191
Bod vzplanutí: 56 °C (133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041419
CAS číslo: 2557-78-0
UN: 3071
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR