2,3,5,6-Tetrafluorophenol ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A12686.06
A12686.14
A12686.06EA
1300
CZK
A12686.06
A12686.14
2,3,5,6-Tetrafluorophenol ≥97%
2,3,5,6-Tetrafluorophenol
Purity: 97%
CAS No.: 769-39-1
Molecular Formula: C6H2F4O
Formula Weight: 166.07
Flash Point: 79°(174°F)
Odour: Phenol-like
Boiling Point: 141-143°
Beilstein Reference: 1911548
UN No.: 3261
MDL No.: MFCD00002157
Vzorec:
C₆H₂F₄O MW: 166,07 g/mol Bod varu: 141…143 °C Bod tání: 28…30 °C Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002157 CAS číslo: 769-39-1 EINECS: 212-209-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...