2,5-Difluorophenol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A18744.06
A18744.14
A18744.06EA
1150
CZK
A18744.06
A18744.14
2,5-Difluorophenol ≥97%
2,5-Difluorophenol
Purity: 97%
CAS No.: 2713-31-7
Molecular Formula: C6H4F2O
Formula Weight: 130.09
Flash Point: 53°(127°F)
Odour: Phenol-like
Beilstein Reference: 2081076
UN No.: 2921
MDL No.: MFCD00042501
Vzorec:
C₆H₄F₂O Bod varu: 166 °C (1013 hPa) Bod tání: 36…40 °C Bod vzplanutí: 53 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042501 CAS číslo: 2713-31-7 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...