Tisk…

Sangerova reagencie ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene

A11871.14 A11871.06 A11871.22
A11871.14EA 2850 CZK
A11871.14 A11871.06 A11871.22
Sangerova reagencie ≥99%
Sangerova reagencie

Purity: 99% CAS No.: 70-34-8 Molecular Formula: C6H3FN2O4 Formula Weight: 186.10 Flash Point: 110°(230°F) Refractive Index: 1.5690 Melting Point: 24-30° Boiling Point: 296° Beilstein Reference: 398632 Merck Reference: 14,4172 UN No.: 2811 MDL No.: MFCD00007056

Vzorec: C₆H₃FN₂O₄
MW: 186.1
Bod varu: 296 °C
Bod tání: 24…30 °C
Hustota: 1.482
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007056
CAS číslo: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1 III
Merck Index: 13,04199

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR