1-Fluoro-3-nitrobenzene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-Fluoronitrobenzene
B25553.14
B25553.22
B25553.14EA
2600
CZK
B25553.14
B25553.22
1-Fluoro-3-nitrobenzene ≥98%
1-Fluoro-3-nitrobenzene
Purity: 98+%
CAS No.: 402-67-5
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Flash Point: 76°(168°F)
Refractive Index: 1.5250
Melting Point: 2°
Boiling Point: 204-206°
Beilstein Reference: 1862210
UN No.: 2810
MDL No.: MFCD00007196
Vzorec:
C₆H₄FNO₂ Bod varu: 204…206 °C Bod tání: 2 °C Hustota: 1,325 Bod vzplanutí: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007196 CAS číslo: 402-67-5 EINECS: 206-953-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...