Tisk…

1,3-Difluoro-4-nitrobenzene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene

A13249.14 A13249.22 A13249.36
A13249.14EA 903 CZK
A13249.14 A13249.22 A13249.36
1,3-Difluoro-4-nitrobenzene ≥98%
1,3-Difluoro-4-nitrobenzene

Purity: 98% CAS No.: 446-35-5 Molecular Formula: C6H3F2NO2 Formula Weight: 159.09 Flash Point: 90°(194°F) Refractive Index: 1.5130 Melting Point: 9-10° Boiling Point: 203-204° Beilstein Reference: 2048034 UN No.: 2810 MDL No.: MFCD00007050

Vzorec: C₆H₃F₂NO₂
MW: 159.09
Bod varu: 203…204 °C
Bod tání: 9…10 °C
Hustota: 1.4571
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007050
CAS číslo: 446-35-5
EINECS: 207-167-0
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR