4-Amino-2,3,5,6-tetrafluorobenzoic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Amino-2,3,5,6-tetrafluorobenzoic Acid
L20128.03
L20128.06
L20128.03EA
1540
CZK
L20128.03
L20128.06
4-Amino-2,3,5,6-tetrafluorobenzoic acid 98%
4-Amino-2,3,5,6-tetrafluorobenzoic acid
Purity: 98%
CAS No.: 944-43-4
Molecular Formula: C7H3F4NO2
Formula Weight: 209.11
Melting Point: 182-184°
Beilstein Reference: 2121117
Vzorec:
C₇H₃F₄NO₂ MW: 209,1 g/mol Bod tání: 182…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007647 CAS číslo: 944-43-4 EINECS: 213-409-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...