Tisk…

6-Fluoroanthranilic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Amino-6-fluorobenzoic acid

A10919.03 A10919.06 A10919.14
A10919.03EA 1170 CZK
A10919.03 A10919.06 A10919.14
6-Fluoroanthranilic acid 98%
6-Fluoroanthranilic acid

Purity: 98% CAS No.: 434-76-4 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Melting Point: 165-167° Beilstein Reference: 3540926

Vzorec: C₇H₆FNO₂
MW: 155.13
Bod varu: 305
Bod tání: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00067781
CAS číslo: 434-76-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR