Tisk…

3-Fluorobenzoic acid ≥99%

Dodavatel: Thermo Scientific
Varování

A16511.14 A16511.22
A16511.14EA 2040 CZK
A16511.14 A16511.22
3-Fluorobenzoic acid ≥99%
3-Fluorobenzoic acid

Purity: 99% CAS No.: 455-38-9 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 122-124° Beilstein Reference: 1906920

Vzorec: C₇H₅FO₂
MW: 140,11 g/mol
Bod varu: 251 °C (1013 hPa)
Bod tání: 122…124 °C
Hustota: 1,474
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS číslo: 455-38-9
EINECS: 207-248-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR