Tisk…

2,4,6-Trifluorobenzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

A19758.03 A19758.06
A19758.03EA 1960 CZK
A19758.03 A19758.06
2,4,6-Trifluorobenzoic acid ≥98%
2,4,6-Trifluorobenzoic acid

Purity: 98% CAS No.: 28314-80-9 Molecular Formula: C7H3F3O2 Formula Weight: 176.10 Melting Point: 140-142° Beilstein Reference: 1958300

Vzorec: C₇H₃F₃O₂
MW: 176,09 g/mol
Bod varu: 237 °C (1013 hPa)
Bod tání: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00042398
CAS číslo: 28314-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR