2,3,4-Trifluorobenzoic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B20696.06
B20696.14
B20696.06EA
3470
CZK
B20696.06
B20696.14
2,3,4-Trifluorobenzoic acid ≥98%
2,3,4-Trifluorobenzoic acid
Purity: 98%
CAS No.: 61079-72-9
Molecular Formula: C7H3F3O2
Formula Weight: 176.10
Melting Point: 140-142°
Beilstein Reference: 7476020
Vzorec:
C₇H₃F₃O₂ Bod varu: 237 °C (1013 hPa) Bod tání: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061232 CAS číslo: 61079-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...