Tisk…

Pentafluorobenzonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,3,4,5,6-Pentafluorobenzonitrile

A13575.06 A13575.14 A13575.22
A13575.06EA 893 CZK
A13575.06 A13575.14 A13575.22
Pentafluorobenzonitrile ≥98%
Pentafluorobenzonitrile

Purity: 98% CAS No.: 773-82-0 Molecular Formula: C7F5N Formula Weight: 193.07 Flash Point: 29°(84°F) Refractive Index: 1.4425 Melting Point: 1-2° Boiling Point: 161-162° Beilstein Reference: 1913453 UN No.: 1992 MDL No.: MFCD00001775

Vzorec: C₇F₅N
Bod varu: 161…162 °C
Bod tání: 1…2 °C
Hustota: 1,566
Bod vzplanutí: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001775
CAS číslo: 773-82-0
EINECS: 212-259-9
UN: 1992
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR