3-Fluorobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B23426.06
B23426.14
B23426.22
B23426.06EA
2050
CZK
B23426.06
B23426.14
B23426.22
3-Fluorobenzonitrile ≥98%
3-Fluorobenzonitrile
Purity: 98%
CAS No.: 403-54-3
Molecular Formula: C7H4FN
Formula Weight: 121.11
Flash Point: 67°(152°F)
Refractive Index: 1.5055
Odour: Almond-like
Melting Point: -16°
Boiling Point: 183°
Beilstein Reference: 1858941
UN No.: 3276
MDL No.: MFCD00001797
Vzorec:
C₇H₄FN Bod varu: 183 °C Bod tání: –16 °C Hustota: 1,140 Bod vzplanutí: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001797 CAS číslo: 403-54-3 EINECS: 206-963-5 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...