Tisk…

2-Fluorobenzonitrile ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: o-Fluorobenzonitrile

A14375.18 A14375.09 A14375.22 A14375.30
A14375.18EA 4530 CZK
A14375.18 A14375.09 A14375.22 A14375.30
2-Fluorobenzonitrile ≥99%
2-Fluorobenzonitrile

Purity: 99% CAS No.: 394-47-8 Molecular Formula: C7H4FN Formula Weight: 121.11 Flash Point: 73°(163°F) Refractive Index: 1.5070 Odour: Almond-like Boiling Point: 196-198° Beilstein Reference: 2042184 MDL No.: MFCD00001773

Vzorec: C₇H₄FN
Bod varu: 196…198 °C
Bod tání: 35…37 °C
Hustota: 1,138
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001773
CAS číslo: 394-47-8
EINECS: 206-897-7
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR