Tisk…

2,6-Difluorobenzonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A10454.09 A10454.18 A10454.30
A10454.09EA 865 CZK
A10454.09 A10454.18 A10454.30
2,6-Difluorobenzonitrile ≥98%
2,6-Difluorobenzonitrile

Purity: 98% CAS No.: 1897-52-5 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 80°(176°F) Refractive Index: 1.4875 Odour: Almond-like Melting Point: 28-32° Boiling Point: 197-198° Beilstein Reference: 2045292 UN No.: 3439 MDL No.: MFCD00001778

Vzorec: C₇H₃F₂N
MW: 139,1 g/mol
Bod varu: 197…198 °C
Bod tání: 28…32 °C
Hustota: 1,246
Bod vzplanutí: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001778
CAS číslo: 1897-52-5
EINECS: 217-589-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR