2,3-Difluorobenzonitrile ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B20811.03
B20811.06
B20811.03EA
954
CZK
B20811.03
B20811.06
2,3-Difluorobenzonitrile ≥99%
2,3-Difluorobenzonitrile
Purity: 99%
CAS No.: 21524-39-0
Molecular Formula: C7H3F2N
Formula Weight: 139.11
Flash Point: 75°(167°F)
Refractive Index: 1.4882
Odour: Almond-like
Boiling Point: 187-188°
Beilstein Reference: 2085621
UN No.: 3276
MDL No.: MFCD00009976
Vzorec:
C₇H₃F₂N Bod varu: 187…188 °C Hustota: 1,254 Bod vzplanutí: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009976 CAS číslo: 21524-39-0 EINECS: 244-418-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...