1,3-Difluorobenzene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A14387.09
A14387.18
A14387.30
A14387.09EA
1230
CZK
A14387.09
A14387.18
A14387.30
1,3-Difluorobenzene ≥99%
1,3-Difluorobenzene
Purity: 99%
CAS No.: 372-18-9
Molecular Formula: C6H4F2
Formula Weight: 114.09
Flash Point: 2°(36°F)
Refractive Index: 1.4380
Odour: Characteristic
Melting Point: -59°
Boiling Point: 82-83°
Beilstein Reference: 1904537
UN No.: 1993
MDL No.: MFCD00000327
Vzorec:
C₆H₄F₂ MW: 114,09 g/mol Bod varu: 82…83 °C Bod tání: –59 °C Hustota: 1,160 Bod vzplanutí: 2 °C (36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000327 CAS číslo: 372-18-9 EINECS: 206-746-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...