Tisk…

1,2,4-Trifluorobenzene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11424.06 A11424.14 A11424.22
A11424.06EA 543 CZK
A11424.06 A11424.14 A11424.22
1,2,4-Trifluorobenzene ≥98%
1,2,4-Trifluorobenzene

Purity: 98+% CAS No.: 367-23-7 Molecular Formula: C6H3F3 Formula Weight: 132.09 Flash Point: 4°(39°F) Refractive Index: 1.4230 Boiling Point: 89-91° Beilstein Reference: 2042865 UN No.: 2387 MDL No.: MFCD00000305

Vzorec: C₆H₃F₃
Bod varu: 89…91 °C
Bod tání: –35 °C
Hustota: 1,264
Bod vzplanutí: 4 °C (39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000305
CAS číslo: 367-23-7
EINECS: 206-684-9
UN: 2387
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR