2,4,5-Trichlorothiophenol ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Mercapto-2,4,5-trichlorobenzene, 2,4,5-Trichlorothiophenol, 2,4,5-Trichlorobenzenethiol, 2,4,5-Trichlorophenyl Mercaptan
L03324.06
L03324.03
L03324.06EA
5010
CZK
L03324.06
L03324.03
2,4,5-Trichlorothiophenol ≥97%
2,4,5-Trichlorothiophenol
Purity: 97%
CAS No.: 3773-14-6
Molecular Formula: C6H3Cl3S
Formula Weight: 213.51
Odour: Unpleasant
Melting Point: 112-118°
Boiling Point: 153-154°/15mm
Beilstein Reference: 509727
UN No.: 3335
MDL No.: MFCD00014447
Vzorec:
C₆H₃Cl₃S MW: 213,51 g/mol Bod varu: 153…154 °C (15 mmHg) Bod tání: 112…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014447 CAS číslo: 3773-14-6 EINECS: 223-223-7 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...