2-Chlorothiophenol ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
o-Chlorothiophenol, 2-chlorothiophenol, 1-Chloro-2-mercaptobenzene, 2-Chlorobenzenethiol, 2-Chlorobenzene-1-thiol
A12014.06
A12014.14
A12014.30
A12014.06EA
1190
CZK
A12014.06
A12014.14
A12014.30
2-Chlorothiophenol ≥98%
2-Chlorothiophenol
Purity: 98%
CAS No.: 6320-03-2
Molecular Formula: C6H5ClS
Formula Weight: 144.62
Flash Point: 88°(190°F)
Refractive Index: 1.6040
Boiling Point: 108-110°/30mm
Beilstein Reference: 1209665
UN No.: 1760
MDL No.: MFCD00004830
Vzorec:
C₆H₅ClS MW: 144,62 g/mol Bod varu: 108…110 °C (30 mmHg) Hustota: 1,277 Bod vzplanutí: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004830 CAS číslo: 6320-03-2 EINECS: 228-667-5 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...