Tisk…

4-Chloro-3-nitrophenol ≥99%

Dodavatel: Thermo Scientific
Varování

L10108.06 L10108.14
L10108.06EA 1400 CZK
L10108.06 L10108.14
4-Chloro-3-nitrophenol ≥99%
4-Chloro-3-nitrophenol

Purity: 99% CAS No.: 610-78-6 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Odour: Characteristic Melting Point: 124-129° Beilstein Reference: 2094546 MDL No.: MFCD00043546

Vzorec: C₆H₄ClNO₃
MW: 173,56 g/mol
Bod tání: 124…129 °C
Hustota: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00043546
CAS číslo: 610-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR