1-Chloro-3-nitrobenzene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-Chloronitrobenzene
B23423.18
B23423.30
B23423.18EA
992
CZK
B23423.18
B23423.30
1-Chloro-3-nitrobenzene ≥98%
1-Chloro-3-nitrobenzene
Purity: 98%
CAS No.: 121-73-3
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Flash Point: 124°(255°F)
Melting Point: 43-47°
Boiling Point: 235-236°
Beilstein Reference: 1907947
Merck Reference: 14,2151
UN No.: 1578
MDL No.: MFCD00007202
Vzorec:
C₆H₄ClNO₂ Bod varu: 235…236 °C Bod tání: 43…47 °C Hustota: 1,34 Bod vzplanutí: 124 °C (255 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007202 CAS číslo: 121-73-3 EINECS: 204-496-1 UN: 1578 ADR: 6.1,II Merck Index: 14,2151 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...