1-chlor-2-nitrobenzen ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Chloro-2-nitrobenzene, 2-Chloronitrobenzene
A15403.0B
A15403.0I
A15403.30
A15403.0BEA
2110
CZK
A15403.0B
A15403.0I
A15403.30
1-chlor-2-nitrobenzen ≥99%
1-chlor-2-nitrobenzen
Purity: 99%
CAS No.: 88-73-3
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Flash Point: 126°(258°F)
Melting Point: 31-34°
Boiling Point: 245-246°
Beilstein Reference: 509273
Merck Reference: 14,2151
UN No.: 1578
MDL No.: MFCD00007061
Vzorec:
C₆H₄ClNO₂ MW: 157,56 g/mol Bod varu: 245…246 °C Bod tání: 31…34 °C Hustota: 1,348 Bod vzplanutí: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007061 CAS číslo: 88-73-3 EINECS: 201-854-9 UN: 1578 ADR: 6.1,II Merck Index: 14,2151 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...