Tisk…

Dicloran (2,6-dichloro-4-nitroaniline) 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,6-Dichloro-4-nitroaniline

B24283.14 B24283.22 B24283.36
B24283.14EA 762 CZK
B24283.14 B24283.22 B24283.36
Dicloran (2,6-dichloro-4-nitroaniline) 95%
Dicloran (2,6-dichloro-4-nitroaniline)

Purity: 95% CAS No.: 99-30-9 Molecular Formula: C6H4Cl2N2O2 Formula Weight: 207.02 Odour: Recognizable Melting Point: 185-192° Boiling Point: 130°/2mm Beilstein Reference: 1459581 MDL No.: MFCD00007677

Vzorec: C₆H₄Cl₂N₂O₂
MW: 207,02 g/mol
Bod varu: 130 °C (2 mmHg)
Bod tání: 185…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00007677
CAS číslo: 99-30-9
EINECS: 202-746-4
UN: 2237
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR