Tisk…

4-Chliranilin ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12879.30 A12879.0B A12879.0I
A12879.30EA 1090 CZK
A12879.30 A12879.0B A12879.0I
4-Chliranilin ≥98%
4-Chliranilin

Purity: 98% CAS No.: 106-47-8 Molecular Formula: C6H6ClN Formula Weight: 127.57 Flash Point: 120°(248°F) Melting Point: 67-72° Boiling Point: 232° Beilstein Reference: 471359 Merck Reference: 14,2118 UN No.: 2018 MDL No.: MFCD00007835

Vzorec: C₆H₆ClN
MW: 127.57
Bod varu: 232 °C
Bod tání: 67…72 °C
Hustota: 1.430
Bod vzplanutí: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS číslo: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1 II
Merck Index: 13,02136

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR