Tisk…

2-Chloroaniline ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A15758.0C A15758.0E A15758.36
A15758.0CEA 15720 CZK
A15758.0C A15758.0E A15758.36
2-Chloroaniline ≥98%
2-Chloroaniline

Purity: 98+% CAS No.: 95-51-2 Molecular Formula: C6H6ClN Formula Weight: 127.57 Flash Point: 97°(206°F) Refractive Index: 1.5880 Odour: Characteristic Melting Point: -3° Boiling Point: 207-209° Beilstein Reference: 606077 Merck Reference: 14,2118 UN No.: 2019 MDL No.: MFCD00007656

Vzorec: C₆H₆ClN
MW: 127,57 g/mol
Bod varu: 207…209 °C
Bod tání: –3 °C
Hustota: 1,213
Bod vzplanutí: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007656
CAS číslo: 95-51-2
EINECS: 202-426-4
UN: 2019
ADR: 6.1,II
Merck Index: 14,2118

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR