Tisk…

2,4,5-Trichloroaniline ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

B21113.18 B21113.30
B21113.18EA 690 CZK
B21113.18 B21113.30
2,4,5-Trichloroaniline ≥97%
2,4,5-Trichloroaniline

Purity: 97% CAS No.: 636-30-6 Molecular Formula: C6H4Cl3N Formula Weight: 196.46 Flash Point: 175°(347°F) Odour: Characteristic Boiling Point: 269-270° Beilstein Reference: 879091 UN No.: 2018 MDL No.: MFCD00007662

Vzorec: C₆H₄Cl₃N
MW: 196,46 g/mol
Bod varu: 269…270 °C
Bod tání: 92…95 °C
Bod vzplanutí: 175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007662
CAS číslo: 636-30-6
EINECS: 211-254-9
UN: 2018
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR