Tisk…

2-Chloro-1-iodo-4-nitrobenzene ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Chloro-4-iodonitrobenzene

A19340.03 A19340.06 A19340.14
A19340.03EA 920 CZK
A19340.03 A19340.06 A19340.14
2-Chloro-1-iodo-4-nitrobenzene ≥98%
2-Chloro-1-iodo-4-nitrobenzene

Purity: 98% CAS No.: 41252-96-4 Molecular Formula: C6H3ClINO2 Formula Weight: 283.45 Melting Point: 97-101° Beilstein Reference: 2502411

Vzorec: C₆H₃ClINO₂
MW: 283.45
Bod tání: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00024589
CAS číslo: 41252-96-4
EINECS: 255-278-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR