Tisk…

1,2-Dichloro-4-iodobenzene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,4-Dichloroiodobenzene

A13440.09 A13440.18
A13440.09EA 1200 CZK
A13440.09 A13440.18
1,2-Dichloro-4-iodobenzene ≥98%
1,2-Dichloro-4-iodobenzene

Purity: 98% CAS No.: 20555-91-3 Molecular Formula: C6H3Cl2I Formula Weight: 272.90 Refractive Index: 1.6480 Melting Point: 29-31° Boiling Point: 116-118°/12mm Beilstein Reference: 2245136 MDL No.: MFCD00019014

Vzorec: C₆H₃Cl₂I
MW: 272,9 g/mol
Bod varu: 116…118 °C (12 mmHg)
Bod tání: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00019014
CAS číslo: 20555-91-3
EINECS: 243-874-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR